Learn More
Sigma Aldrich Fine Chemicals Biosciences KimaxR heavy-duty wide-mou
Catalog No. 502906144
Encompass
Wall thickness and weight are at least two times greater than that of standard volumetric flasks for durability. Calibrated to contain. Meets ASTM E288 Class A unserialized requirements. Easy pipette access. Supplied with glass stopper.
Catalog No. 50-290-6144 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z5115286EA
Wall thickness and weight are at least two times greater than that of standard volumetric flasks for durability. Calibrated to contain. Meets ASTM E288 Class A unserialized requirements. Easy pipette access. Supplied with glass stopper.