Learn More
Sigma Aldrich Fine Chemicals Biosciences KimaxR weighing bottle Sty
Catalog No. 502905074
Encompass
Designed from federal Specification NNN-B-1493 requirements.Cylindrical tall form. Style I has a closed bottom stopper that fits into an internally ground body.
Catalog No. 50-290-5074 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2749412PAK
Designed from federal Specification NNN-B-1493 requirements.Cylindrical tall form. Style I has a closed bottom stopper that fits into an internally ground body.