Learn More
Kimberly-Clark™ Professional Hard Hats
Catalog No. 17350203
Click to view available options
Color:
Green
Yellow
Size:
6.5 to 8
Standard
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
17-350-203 Yellow 6.5 to 8
17-350-204 Green Standard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-350-203 Supplier Kimberly-Clark™ Professional Supplier No. 14833
Kimberly-Clark™ Professional Hard Hats have a four-point ratchet suspension that adjusts to fit heads from sizes 6.5 to 8
  • Meet ANSI Z89.1 - 2009 standards for impact (as well as Class C, G and E)
  • Smooth dome design with no crevices
  • Comfortable to wear, even for long periods of time, since they are molded from lightweight, dielectric HDPE (high density polyethylene)
  • Features accessory slots for attaching additional personal protective equipment
Specifications
Certifications/Compliance Meets ANSI Z89.1; Class C, G and E
Color Yellow
Material HDPE
Quantity 12/Cs.
Size 6.5 to 8
Suspension Type 4-Point Ratchet