Learn More
Kimberly-Clark™ Professional Jackson Safety™ BC100 Bump Caps
Catalog No. 191501563
Click to view available options
Color:
Blue
Red
White
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-150-1563 Red
19-150-1559 White
19-150-1561 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-150-1563 Supplier Kimberly-Clark™ Professional Supplier No. 14815
This item is not returnable. View return policy
Kimberly-Clark™ Professional Jackson Safety™ BC100 Bump Caps offer protection from minor bumps and scalp lacerations
  • Lightweight for comfort
  • Made of molded HDPE (high-density polyethylene)
  • 4-point injection molded system includes an absorbent brow pad
  • Pinlock sizing system can accommodate head sizes 6.5 to 8.25
  • Can be made compatible with a face shield for additional coverage
  • Replacement sweat bands and suspension inserts available
Specifications
Color Red
Material HDPE
Product Type Low-profile Bump Cap
Quantity 12/Pk.
Size Adjustable (Pin-lock sizing)
Suspension Type 4-Point Injection Molded