Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kimberly-Clark Professional™ Kimtech™ W Aviation Wet Wipe System

Catalog No. 17000247 Shop All Kimberly-Clark Professional Products
Change view
Click to view available options
Dimensions (L x W):
8.6 x 9.25 in. (22 x 23 cm)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
17-000-247 8.6 x 9.25 in. (22 x 23 cm)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-000-247 Supplier Kimberly-Clark Professional™ Supplier No. 28646

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Air-tight, sealed system reduces waste and improves performance by offering more controlled dispensing of disposable, pre-saturated wipes

  • Benefits include reductions in solvent loss from evaporation, reduced waste from wipes drying out, and reduced worker exposure to Volatile Organic Compounds (VOCs)
  • Independent bucket and wipe system, separate from solvent requirements
  • Air-tight, sealed container with gasket
  • Screw on lid with snap-top cover and large lead-wipe reservoir
  • More controlled dispensing and application of solvent versus bucket or decanter systems allowing for:
    • A measured amount of solvent poured in a closed container
    • An entire wipe to be saturated with solvent
    • Write-on color-coded labels
    • Wipe and solvent together in one package with a handle
    • Durable and chemical resistant construction makes the bucket reusable
  • Overall general cleaning, Post paint stripping wiping, Water rinse drying, Solvent wiping, Excess sealant removal, Oil, grease cleaning, Tool wiping

Specifications

Product Type Aviation Wipes Dispenser Bucket
Material Plastic
Autoclavable Not Autoclavable
Disposable Reusable
Dimensions (L x W) 8.6 x 9.25 in. (22 x 23 cm)
Format Dispenser bucket
For Use With (Application) Overall general cleaning, post paint stripping wiping, Water rinse drying, solvent wiping, excess sealant removal, oil, grease cleaning, tool wiping
For Use With (Equipment) WetTask™ refillable wet wiping system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.