Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kimberly-Clark Professional™ Kleenex™ Antibacterial Foam Skin Cleanser

Catalog No. 19014902 Shop All Kimberly-Clark Professional Products
Change view
Click to view available options
Volume (Metric):
1.2 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
19-014-902 1.2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-014-902 Supplier Kimberly-Clark Professional™ Supplier No. 91594
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Daily use mild skin cleanser to kill harmful germs

  • Kills harmful germs such as E. coli, Staphylococcus aureaus and Salmonella
  • Contains 0.5% Triclosan with moisturizers such as Aloe Vera, Glycerin, and Provitamin B5

Specifications

Product Type Anti-bacterial Foam Soap
Color Clear
Composition Contains 0.5% Triclosan with moisturizers such as Aloe Vera, Glycerin, and Provitamin B5
Dimensions (L x W x H) 6.687 x 6.125 x 11.000 in.
For Use With (Application) Kills harmful germs such as E. coli, Staphylococcus aureaus and Salmonella
Volume (Metric) 1.2 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.