Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kimberly-Clark Professional™ Scott™ Essential™ Plus+ Hard Roll Towels
GREENER_CHOICE

Catalog No. 19413022 Shop All Kimberly-Clark Professional Products
Catalog No. 19-413-022 Supplier Kimberly-Clark Professional™ Supplier No. 50606
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Soft feel, premium performance in a roll towel, preferred for hand drying performance. New Zip-Off Top packaging. Absorbs lots of water fast, for superior hand drying. Meets EPA standards with a minimum of 40% post-consumer waste. Smaller case for easier handling and storage.

  • Small 6 roll case size for easy storage (1 roll = 600 feet, 1 case = 3600 feet); Same Kleenex quality, now Scott branded
  • Scott Essential (formerly Kleenex) paper towel rolls give soft feel with the premium performance of commercial paper towels
  • Absorbency Pockets absorb lots of water fast for superior hand drying performance, so guests use fewer paper towels
  • Paper towel rolls: 1.75 in. core size and 7.9 in. roll diameter
  • Compatible with a variety of Kimberly-Clark Professional paper towel dispensers

Specifications

Product Type Towel
Certifications/Compliance EPA
Color White
For Use With (Application) Hand drying performance
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.