Learn More
MSC Kimberly-Clark KleenGuard A20 Coveralls, White, 2X-Large, 24/Case
Catalog No. 5000676034
Encompass
The KleenGuard A20 Breathable Particle Protection Coverall provides essential protection for workers against common contaminants like dirt, grime, and dry particles. It is designed with MICROFORCE™ Barrier SMS fabric, which keeps workers clean and comfortable in environments with light exposure to non-hazardous particulates and liquid sprays.
  • Offers resistance against dry particles, abrasion, and light liquid sprays, ensuring compliance in mildly hazardous environments.
  • Reflex Coverall design provides superior fit and freedom of movement, enhancing worker efficiency and comfort.
  • Constructed with breathable yet strong MICROFORCE™ Barrier fabric, standing up to heavy dust environments.
  • Features an attached hood, elastic back/wrists/ankles, and a zipper front with a storm flap.
  • Passes NFPA 99 for antistatic materials.
Catalog No. 50-006-76034 Supplier MSC Supplier No. 06589659
The KleenGuard A20 Breathable Particle Protection Coverall provides essential protection for workers against common contaminants like dirt, grime, and dry particles. It is designed with MICROFORCE™ Barrier SMS fabric, which keeps workers clean and comfortable in environments with light exposure to non-hazardous particulates and liquid sprays.
  • Offers resistance against dry particles, abrasion, and light liquid sprays, ensuring compliance in mildly hazardous environments.
  • Reflex Coverall design provides superior fit and freedom of movement, enhancing worker efficiency and comfort.
  • Constructed with breathable yet strong MICROFORCE™ Barrier fabric, standing up to heavy dust environments.
  • Features an attached hood, elastic back/wrists/ankles, and a zipper front with a storm flap.
  • Passes NFPA 99 for antistatic materials.