Learn More
Enterprise Technology Solutions Kimberly-Clark Professional Cleaning Towels, Pop-Up Box, 9-1/10"x16-4/5", 118/BX, WE
SDP
Catalog No. 5000831633
Encompass_Preferred
Create a clean work environment with light-duty to medium-duty, general cleaning solutions. General Clean X60 Multi-Task Cleaning Cloths absorb more than two times as much water as rags. They bring the superior absorbing power of HydroKnit technology to a lighter-weight cloth. Designed to outperform laundered rags, these cloths help raise the standard for general-purpose cloths. They don't use adhesives or binders so they'll leave no residue behind. Cloths are reinforced for enhanced strength when wet or dry. Use cloths for a multitude of purposes such as applying thinners and solvents, removing cable lubricant, cleaning adhesive cleaning and removing grease/soils in tight areas. Cleaning towels come in a pop-up box dispenser and are sturdy enough to be used more than once to help reduce waste. They are disposable after you use them a few times.
Catalog No. 50-008-31633 Supplier Enterprise Technology Solutions Supplier No. KCC34790
Create a clean work environment with light-duty to medium-duty, general cleaning solutions. General Clean X60 Multi-Task Cleaning Cloths absorb more than two times as much water as rags. They bring the superior absorbing power of HydroKnit technology to a lighter-weight cloth. Designed to outperform laundered rags, these cloths help raise the standard for general-purpose cloths. They don't use adhesives or binders so they'll leave no residue behind. Cloths are reinforced for enhanced strength when wet or dry. Use cloths for a multitude of purposes such as applying thinners and solvents, removing cable lubricant, cleaning adhesive cleaning and removing grease/soils in tight areas. Cleaning towels come in a pop-up box dispenser and are sturdy enough to be used more than once to help reduce waste. They are disposable after you use them a few times.