Learn More
Enterprise Technology Solutions Kimberly-Clark Professional Coveralls, f/Particle Protection, A20, 3XL, 20/CT, Blue
SDP
Catalog No. 5000828296
Encompass_Preferred
Particle protective coveralls are great for laboratories, decontamination modesty garment, remediation, pesticides and heavy dust environments. Patented Reflex design is 7-1/2 times less likely to rip out than ANSI minimums. It also provides 12 percent more chest room, 6 percent longer body length and 9 percent total yards of fabric. Comfortable shell features a zipper front and elastic on the wrists and ankles. The KleenGuard A20 Breathable Particle Protection Coveralls pass the NFPA 99 criteria for antistatic materials. Patented MicroForce Barrier SMS fabric offers a barrier against dry particulates. Seamless front provides more protection in the primary exposure areas. 3XL size fits bodies from 6'2" to 6'7" tall that weigh 240 to 280 lb.
Catalog No. 50-008-28296 Supplier Enterprise Technology Solutions Supplier No. KCC58506
Particle protective coveralls are great for laboratories, decontamination modesty garment, remediation, pesticides and heavy dust environments. Patented Reflex design is 7-1/2 times less likely to rip out than ANSI minimums. It also provides 12 percent more chest room, 6 percent longer body length and 9 percent total yards of fabric. Comfortable shell features a zipper front and elastic on the wrists and ankles. The KleenGuard A20 Breathable Particle Protection Coveralls pass the NFPA 99 criteria for antistatic materials. Patented MicroForce Barrier SMS fabric offers a barrier against dry particulates. Seamless front provides more protection in the primary exposure areas. 3XL size fits bodies from 6'2" to 6'7" tall that weigh 240 to 280 lb.