Learn More
Enterprise Technology Solutions Kimberly-Clark Professional Coveralls,Zipper Front,Elastic Wrists/Ankles,3XL,25/CT,White
SDP
Catalog No. 5000828295
Encompass_Preferred
Kleenguard A40 Liquid and Particle Protection Coveralls offer your team comfortable clothing to wear as they work. KleenGuard Personal Protective Equipment enables rather than inhibits maximum productivity and safety for you and your employees. Each protective suit comes with a zipper front and elastic wrists and ankles. Coveralls are made of breathable, microporous film to keep out debris, nonhazardous liquid chemicals and dry particulates. Kleenguard A40 material gives your employees a better liquid barrier and a better particulate barrier than Dupont Tyvek. These coveralls pass NFPA 99 for antistatic materials, pass ASTM F1670 testing for penetration of blood and bodily fluids, and meet the ANSI/ISEA 101-1996 sizing standards. Popular industries using this product include aviation, pharmaceutical manufacturing, liquid handling, general manufacturing and so much more.
Catalog No. 50-008-28295 Supplier Enterprise Technology Solutions Supplier No. KCC44316
Kleenguard A40 Liquid and Particle Protection Coveralls offer your team comfortable clothing to wear as they work. KleenGuard Personal Protective Equipment enables rather than inhibits maximum productivity and safety for you and your employees. Each protective suit comes with a zipper front and elastic wrists and ankles. Coveralls are made of breathable, microporous film to keep out debris, nonhazardous liquid chemicals and dry particulates. Kleenguard A40 material gives your employees a better liquid barrier and a better particulate barrier than Dupont Tyvek. These coveralls pass NFPA 99 for antistatic materials, pass ASTM F1670 testing for penetration of blood and bodily fluids, and meet the ANSI/ISEA 101-1996 sizing standards. Popular industries using this product include aviation, pharmaceutical manufacturing, liquid handling, general manufacturing and so much more.