Learn More
Enterprise Technology Solutions Kimberly-Clark Professional Exam Gloves,Nitrile,Textured Fingertips,XSmall,200/BX,PE
SDP
Supplier:  Enterprise Technology Solutions KCC62825
Encompass_Preferred
Kimtech Vista Nitrile Exam Gloves reduce waste while providing excellent glove dexterity, strength and protection. They are fully recyclable through the RightCycle program so your lab gloves won't end up in landfills. They have a strong, wet and dry grip that reduces the potential for slips. Textured fingertips give your lab technicians improved grip and dexterity in their fine-motor skills. Using these nitrile gloves help cut down on waste and enhances performance in pharma labs, university labs and more.
Catalog No. 50-008-28301