Learn More
Kimberly Clark Professional Kimberly-Clark Professional™ ICON™ Coreless Standard Roll Toilet Paper Dispenser 4 Roll
Catalog No. 17103054 Shop All Kimberly-Clark Professional Products
Change view
Click to view available options
Mounting:
Wall Mount
Catalog No. Mounting
17-103-054 Wall Mount
1 options
Catalog No. 17-103-054 Supplier Kimberly Clark Professional Supplier No. 53705
  • $126.00
    $100.65 / Case of 1
    Save $25.35
    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Made of durable plastic and holds four tissue rolls in a side-by-side configuration. It delivers an exceptional experience and provides consumption control and waste reduction. The dispenser is manually operated and opens using either the keyless push button option or the included key.
The Kimberly-Clark Professional ICON™ dispensing system is our newest industry-leading innovation, changing the face of the restroom forever.
  • Featuring first-of-a-kind versatility, the ICON dispenser effortlessly enhances the look of the restroom with an exclusive line of designer faceplates to help reimagine the restroom.
  • The ICON 4 Roll Toilet Paper Dispenser provides state of the art solutions to maximize productivity.
  • The ICON dispenser is made of durable plastic and holds four tissue rolls in a side-by-side configuration.
  • Designed with the contemporary, stylish profile, it delivers an exceptional experience and provides consumption control and waste reduction.
  • The dispenser is manually operated and opens using either the keyless push button option or the included key.
  • The rolls are placed on two tissue roll holders (known as H mandrels), designed for coreless or small core toilet paper.
  • The rolls are placed on each end of the H mandrel and with the door closed, the four rolls are visible in the dispenser.
  • They are easily accessible through the front with no need to open the dispenser to access another roll.
  • When properly installed and when using recommended product, the dispenser meets the ADA Standards for Accessible Design.
  • These gorgeous dispensers feature our interchangeable faceplate technology and complement the other ICON paper towel and skin care dispensers in the collection.
  • The toilet paper rolls that fit this ICON dispenser are Scott™ and Cottonelle™ Toilet Paper SKUS: 04007; 07001; 47305; 53862.
Specifications
Product Type Toilet Paper Dispenser
Color Silver
Holds Bathroom Tissue
Material Plastic, Metal
Mounting Wall Mount
No. of Compartments 1
Product Style Front-loading with Opening