Learn More
Kimberly-Clark Professional™ Kleenex™ General™ Gentle Skin Cleansers
Shop All Kimberly-Clark Professional ProductsDescription
- Mild, all purpose moisturising skin cleanser
- Ideal for public facilities including hospitality, office buildings, hospitals, restaurants, etc.
- pH balanced resulting in healthy hands
- Pleasant floral fragrance
Specifications
Specifications
| Product Type | Skin Cleanser |
| Color | Pink |
| Form | Foam |
| Boiling Point | >100°C |
| Composition | Sodium Laureth Sulfate 5 to 10%, Cocamidopropyl Betaine 3 to 7%, non-hazardous ingredients 83 to 92% |
| Disposable | Single-use |
| For Use With (Application) | Used to cleanse or remove light soil on Gentle Skin |
| For Use With (Equipment) | Kimberly-Clark Professional™ Cassette Skin Care Dispenser |
| Latex Free | Yes |
| Packaging | Dispenser |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.