Learn More
MSC Kimberly-Clark Professional N95 Particulate Respirator, Valved, Molded Cup, 10/Box
Catalog No. 5000781849
Encompass
The KleenGuard 3300 Series Particulate Respirator provides at least 95% filtration efficiency for 0.3 micron particles. Designed for comfort and a secure fit, this N95 respirator is suitable for various industrial applications and includes an exhalation valve.
  • At least 95% filtration efficiency for 0.3 micron particles
  • Contoured molded cup design
  • Secure fit with welded head straps and nose clip
  • Exhalation valve for reduced heat and easier breathing
  • Soft foam nose seal
  • Not made with natural rubber latex
  • Meets NIOSH N95 standards
  • Standard size
Catalog No. 50-007-81849 Supplier MSC Supplier No. 29630423
The KleenGuard 3300 Series Particulate Respirator provides at least 95% filtration efficiency for 0.3 micron particles. Designed for comfort and a secure fit, this N95 respirator is suitable for various industrial applications and includes an exhalation valve.
  • At least 95% filtration efficiency for 0.3 micron particles
  • Contoured molded cup design
  • Secure fit with welded head straps and nose clip
  • Exhalation valve for reduced heat and easier breathing
  • Soft foam nose seal
  • Not made with natural rubber latex
  • Meets NIOSH N95 standards
  • Standard size