Learn More
Enterprise Technology Solutions Kimberly-Clark Professional Safety Glasses,Nemesis,Polarized Smoke/GY Frame
SDP
Catalog No. 5000828297
Encompass_Preferred
V30 Nemesis Safety Glasses offer stylish good looks and a durable, lightweight construction that's great for protection both at work and home. KleenGuard Personal Protective Equipment enables rather than inhibits maximum productivity and safety for you and your employees. Polycarbonate lenses provide 99.9 percent UVA/UVB/UVC protection to help prevent eye damage like cataracts, retinal damage and other conditions that can cause temporary vision loss. Polarized smoke lenses meet EN 1836 standard for polarization. Wraparound frame has a base curve of 8 that extends the range of peripheral protection as well as soft-touch temples for comfort. Unisex safety glasses have an extremely flexible, patented nose piece to channel sweat away from your eyes and help prevent slippage down the nose. Each pair of safety glasses comes with a neck cord to keep close and protect your eyes at a moment's notice. Safety eyewear complies with the ANSI/ISEA Z87.1 plus standard for personal eye protection.
Catalog No. 50-008-28297 Supplier Enterprise Technology Solutions Supplier No. KCC28635
V30 Nemesis Safety Glasses offer stylish good looks and a durable, lightweight construction that's great for protection both at work and home. KleenGuard Personal Protective Equipment enables rather than inhibits maximum productivity and safety for you and your employees. Polycarbonate lenses provide 99.9 percent UVA/UVB/UVC protection to help prevent eye damage like cataracts, retinal damage and other conditions that can cause temporary vision loss. Polarized smoke lenses meet EN 1836 standard for polarization. Wraparound frame has a base curve of 8 that extends the range of peripheral protection as well as soft-touch temples for comfort. Unisex safety glasses have an extremely flexible, patented nose piece to channel sweat away from your eyes and help prevent slippage down the nose. Each pair of safety glasses comes with a neck cord to keep close and protect your eyes at a moment's notice. Safety eyewear complies with the ANSI/ISEA Z87.1 plus standard for personal eye protection.