Learn More
Kimberly-Clark™ Professional Smith & Wesson™ Phantom Safety Eyewear
Description
- Seven-base spherical lens
- Ribbed, non-slip rubber nose pad
- Straight-back bayonet design temples eliminate mastoid bone pressure
- UV protection
Compliance
Meets ANSI Z87.1 - 2010 High Velocity Industrial Standards
Specifications
Specifications
| Certifications/Compliance | Exceeds ANSI Z87.1 - 2010 High Velocity Industrial Standards |
| Lens Tint | Orange |
| Frame Color | Copper |
| Temple Style | Straight back bayonet design |