Learn More
Enterprise Technology Solutions Kimberly-Clark Professional Task Wipes,Delicate,1 Ply,4.39"x8.2",286/PK,30PK/CT, White
SDP
Catalog No. 5000811306
Encompass_Preferred
When your work requires delicate, light-duty wipers to handle precise tasks, these Kimwipes Delicate Task Wipers are a perfect choice. With a legacy of more than 60 years of being the go-to wipe for cleaning surfaces, parts and instruments in labs, laboratory lenses and medical offices, these single-ply wipes easily clean liquids, dust and small particles. Antistatic dispensing design reduces electrostatic discharge. Medical wipes are an efficient, waste-reducing essential thanks to one-at-a-time dispensing.
Catalog No. 50-008-11306 Supplier Enterprise Technology Solutions Supplier No. KCC34120
When your work requires delicate, light-duty wipers to handle precise tasks, these Kimwipes Delicate Task Wipers are a perfect choice. With a legacy of more than 60 years of being the go-to wipe for cleaning surfaces, parts and instruments in labs, laboratory lenses and medical offices, these single-ply wipes easily clean liquids, dust and small particles. Antistatic dispensing design reduces electrostatic discharge. Medical wipes are an efficient, waste-reducing essential thanks to one-at-a-time dispensing.