Learn More
Enterprise Technology Solutions Kimberly-Clark Professional Wipers,Dry,Dispos.,f/Cleanroom ISO 5,Poly/Rayon,100/PK,5/CT,WE
SDP
Catalog No. 5000830838
Encompass_Preferred
Pure W5 Dry Wipers deliver a delicate cleaning solution for sensitive environments. Disposable wipers assist with critical tasks in clean rooms (approved ISO Class 5 or higher) and pharmaceutical, biotechnology and medical-device industries. 50/50 polyester-and-rayon blend gives wipers cloth-like feel. High-absorbency wipes come in anti-stat, double-bagged packaging.
Catalog No. 50-008-30838 Supplier Enterprise Technology Solutions Supplier No. KCC06179
Pure W5 Dry Wipers deliver a delicate cleaning solution for sensitive environments. Disposable wipers assist with critical tasks in clean rooms (approved ISO Class 5 or higher) and pharmaceutical, biotechnology and medical-device industries. 50/50 polyester-and-rayon blend gives wipers cloth-like feel. High-absorbency wipes come in anti-stat, double-bagged packaging.