Learn More
Enterprise Technology Solutions Kimberly-Clark Professional Wypall X70 Wipers, pop-Up Box, 96 Shts/BX, 10BX/CT, WE
SDP
Catalog No. 5000830542
Encompass_Preferred
Maximize performance while minimizing time, effort and product waste by using WypAll PowerClean X70 Medium-Duty Cloths. High-tech, HydroKnit material absorbs quickly. Premium industrial cloths are made of soft pulp fibers bonded to a polypropylene base sheet for absorbency and tear-resistance so they remove dirt, oil, grime and solvents in a variety of industrial and manufacturing industries just like laundered shop towels or textile shop rags. Durable cloths come in a convenient, Pop-Up Box that dispenses one at a time as an alternative to a box of rags. They are so sturdy that they can often be used more than once. After you've used them a few times, throw away the disposable cloths. They are designed to clean surfaces and tools, making them a must-have for the HVAC, automotive, manufacturing and industrial industries.
Catalog No. 50-008-30542 Supplier Enterprise Technology Solutions Supplier No. KCC41455CT
Maximize performance while minimizing time, effort and product waste by using WypAll PowerClean X70 Medium-Duty Cloths. High-tech, HydroKnit material absorbs quickly. Premium industrial cloths are made of soft pulp fibers bonded to a polypropylene base sheet for absorbency and tear-resistance so they remove dirt, oil, grime and solvents in a variety of industrial and manufacturing industries just like laundered shop towels or textile shop rags. Durable cloths come in a convenient, Pop-Up Box that dispenses one at a time as an alternative to a box of rags. They are so sturdy that they can often be used more than once. After you've used them a few times, throw away the disposable cloths. They are designed to clean surfaces and tools, making them a must-have for the HVAC, automotive, manufacturing and industrial industries.