Learn More
MSC Kimberly-Clark Wrap-a-Round Tape Measure, 10 in. Pipe Dia, 72 in.
Catalog No. 5000673973
Encompass
These wrap-around tape measures fit around a pipe to measure its circumference. The tape also provides a straight edge that wraps around the pipe for marking cut lines. Tape measures are also marked with a tangent chart that helps mark the pipe for angled cuts.
  • Service temperature of up to 350°F
  • Asbestos free
  • Scale in inches
  • Pitch chart
  • Tangent chart and other markings
Catalog No. 50-006-73973 Supplier MSC Supplier No. 65579146
These wrap-around tape measures fit around a pipe to measure its circumference. The tape also provides a straight edge that wraps around the pipe for marking cut lines. Tape measures are also marked with a tangent chart that helps mark the pipe for angled cuts.
  • Service temperature of up to 350°F
  • Asbestos free
  • Scale in inches
  • Pitch chart
  • Tangent chart and other markings