Learn More
Thermo Scientific™ Kimble 11mm Aluminum-Seal LOVial Larger Opening Crimp Top Vials
Shop All Thermo Scientific ProductsDescription
- 6mm I. D. opening—40% larger than standard opening
- Vial capacity, 1.8mL
- Ekonical™ vial capacity, 0.1mL
Specifications
Specifications
| Material | Silica Glass |
| Volume (Metric) | 0.1 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.