Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ Black with Cone-Shaped Liner Phenolic Caps/Closures
SDP

Catalog No. 03340207 Shop All DWK Life Sciences Products
Change view
Click to view available options
Closure Size:
13-425
15-425
18-400
22-400
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Size
03-340-207 15-425
03-340-206 13-425
03-340-208 18-400
03-340-210 22-400
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 03-340-207 Supplier DWK Life Sciences Supplier No. 7380915425
Add to Cart
Add to Cart

Black caps and closures offer excellent sample storage and re-sealing properties

  • Phenolic
  • Cone-shaped liner for two-part seal
  • Superior torque retention

TRUSTED_SUSTAINABILITY

Specifications

Material (Liner) Polyethylene
Color Black
Material Phenolic
For Use With (Application) Excellent sample storage and re-sealing properties
Closure Size 15-425
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.