Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ 20mL Polypropylene Scintillation Vial, Closures Packed Separately
SDP

Catalog No. 0333728 Shop All DWK Life Sciences Products
Catalog No. 03-337-28 Supplier DWK Life Sciences Supplier No. 5851120

missing translation for 'validMainframeMsg'

Foamed Polyethylene cap liner

  • Vials are packaged in cellular trays of 100 pieces
  • Trays are shrink wrapped to preserve cleanliness
  • Closures are packed in polyethylene bags of 100 pieces in separate tray

TRUSTED_SUSTAINABILITY

Specifications

Material (Liner) Foamed Polyethylene
Material HDPE
Volume (Metric) 20 mL
Packaging Type Cellular Trays
Volume (English) 0.67 oz.
Closure Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.