Learn More
DWK Life Sciences™ Kimble™ KIMAX™ Vigreux Distillation Column with Drip Tip, 200mm, 19/22
SDP
Catalog No. 1130149 Shop All DWK Life Sciences Products
Change view
Click to view available options
Standard Taper Joint:
19/22
Catalog No. Standard Taper Joint
11-301-49 19/22
1 options
Catalog No. 11-301-49 Supplier DWK Life Sciences™ Supplier No. 21802200
Add to Cart
Add to Cart
KIMAX™ column has a Standard Taper outer joint at the top and a Standard Taper inner joint at the bottom.
  • Drip tip is provided at the lower end
  • Inner tube has twelve sets of rings of indentations spaced approximately 14mm apart
  • Sets are alternately horizontal and slanted downward
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
TRUSTED_SUSTAINABILITY
Specifications
Includes Drip tip, supplied with jacket to make column a condenser
Type Vigreux Column
Certifications/Compliance USP Type I, ASTM E438, Type I, Class A
Standard Taper Joint 19/22