Learn More
DWK Life Sciences KIMBLE™ KIMAX™ Narrow-Mouth Solution Bottles with Color-Coded PTFE Standard Taper Flathead Stoppers
Shop All DWK Life Sciences Products

Description
Includes
Stoppers
- KimCote™ plastic coating prevents and contains spills, reducing hazards from shattered glass or leakage of toxic or corrosive chemicals
- Ultra-clear borosilicate glass bottles for improved sample viewing
- Non-slip surface, wet or dry
- Autoclavable and chemical resistant
- Narrow-mouth design increases mechanical strength
- Standard Taper ground necks accept flathead color-coded PTFE stoppers
- Stoppers color-coded for easy identification
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
- Replacement stoppers available separately (Part No. 41941)
Compliance
Conforms to ASTM™ E438, Type 1, Class A glass
Specifications
Specifications
| Capacity | 3.3 oz., 100 mL |
| Capacity (Metric) | 100 mL |
| Coating | Plastic |
| Color | Red |
| Diameter (English) | 2 in. |
| Material | Borosilicate Glass |
| Sterility | Non-sterile |
| Closure Color | Red |
| Closure Included | Yes |
| Graduated | No |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.