Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences KIMBLE™ KIMAX™ Narrow-Mouth Solution Bottles with Color-Coded PTFE Standard Taper Flathead Stoppers
GSA_VASDP

Catalog No. 12141316 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
2000 mL
Closure Color:
Blue
Gray
Green
Red
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Color
12-141-316 250 mL Gray
12-141-315 100 mL Red
12-141-317 500 mL Blue
12-141-318 1000 mL Green
12-141-319 2000 mL Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 12-141-316 Supplier DWK Life Sciences Supplier No. KC15097250
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Stoppers

KIMBLE™ KIMAX™ Narrow-Mouth Solution Bottles with Color-Coded PTFE Standard Taper Flathead Stoppers are designed for storage and media preparation. KimCote™ plastic coating provides spill containment and handling protection, while color-coded PTFE stoppers enable easy identification.

  • KimCote™ plastic coating prevents and contains spills, reducing hazards from shattered glass or leakage of toxic or corrosive chemicals
  • Ultra-clear borosilicate glass bottles for improved sample viewing
  • Non-slip surface, wet or dry
  • Autoclavable and chemical resistant
  • Narrow-mouth design increases mechanical strength
  • Standard Taper ground necks accept flathead color-coded PTFE stoppers
  • Stoppers color-coded for easy identification
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
  • Replacement stoppers available separately (Part No. 41941)

Compliance

Conforms to ASTM™ E438, Type 1, Class A glass

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Capacity 8 oz., 250 mL
Capacity (Metric) 250 mL
Coating Plastic
Color Gray
Diameter (English) 2.8 in.
Material Borosilicate Glass
Sterility Non-sterile
Closure Color Gray
Closure Included Yes
Graduated No
Mouth Narrow
Height (Metric) 145 mm
Height (English) 5.7 in.
Closure Material PTFE
Closure Size 19 mm
Diameter (Metric) 72 mm
Closure Type Flat Stopper
Autoclavable Autoclavable
Capacity (English) 8 oz.
Shape Round
Certifications/Compliance ASTM E438, Type I, Class A
Disposable Reusable
Product Type Safety-coated Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.