Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences KIMBLE™ KONTES™ Antibiotic Moisture Content Flasks
GSA_VASDP

Catalog No. K8817000000 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
2.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
K881700-0000 2.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K881700-0000 Supplier DWK Life Sciences Supplier No. 8817000000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KIMBLE™ KONTES™ Antibiotic Moisture Content Flasks are designed for determining moisture loss on drying in tablets, troches, or capsules. Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.

For determining moisture loss on drying in tablets, troches, or capsules

  • Designed for determining moisture loss on drying in tablets, troches, or capsules
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
  • Sample is weighed, transferred to the flask and placed in a vacuum oven
  • Drying is performed at 60°C and a pressure of 5 mm or less of mercury for three hours
  • Percent moisture loss is then calculated from the dried sample weight

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 2.2 mL
Material Borosilicate Glass
For Use With (Application) For determining moisture loss on drying in tablets, troches, or capsules
Product Type Flask
Standard Taper Joint 10/10
Autoclavable Autoclavable
Disposable Reusable
Graduated No
Mouth Style Joint
Sterility Non-sterile
Bottom Shape Flat
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.