Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences™ Kimble™ Kontes™ Distillation Apparatus Components: 100mL Flask
SDP

Catalog No. K6050301524 Shop All DWK Life Sciences Products
Change view
Click to view available options
Standard Taper Joint:
24/40
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint
K605030-1524 24/40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K605030-1524 Supplier DWK Life Sciences™ Supplier No. 6050301524
Add to Cart
Add to Cart

Round Bottom Distilling Flask with Standard Taper Joint Sidearm is a component of Ammonia Nitrogen Distillation Apparatus 552500-1000.

  • Jointed Distilling Flask
  • Standard Taper 24/40
  • 1000mL capacity
  • Case of 1
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
TRUSTED_SUSTAINABILITY

Specifications

Includes Standard taper joint sidearm
Type Distilling Flask
Material Borosilicate Glass
Capacity 33.81 oz., 1000mL
Certifications/Compliance USP Type I and ASTM E438, Type I, Class A
Capacity (Metric) 1000mL
For Use With (Equipment) Distillation apparatus
Standard Taper Joint 24/40
Capacity (English) 33.81 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.