Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ Kontes™ KIMAX™ KimCote™ Heavy-Duty Serum Bottles
SDP

Catalog No. p-3827191 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
4000 mL
9000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
12-141-307 4000 mL
12-141-308 9000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-141-307 Supplier DWK Life Sciences Supplier No. KC149604
Only null left
Add to Cart
Add to Cart

Plastic-coated for added safety

  • KimCote™ plastic coating for added safety in the event of breakage
  • Autoclavable
  • Ultra-clear and extremely durable
  • Resistant to many common laboratory chemicals
  • Unique texture provides non-slip handling, wet or dry

TRUSTED_SUSTAINABILITY

Specifications

Capacity 135.2 oz., 4000mL
Capacity (Metric) 4000 mL
Capacity (English) 135.2 oz.
Color Clear
Diameter (English) 6.57 in.
Material Glass
Height (Metric) 310 mm
Height (English) 12.2 in.
Closure Material Rubber
Closure Size No. 8
Diameter (Metric) 167 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.