Learn More
DWK Life Sciences Kimble™ Kontes™ Kuderna-Danish Evaporative Concentrator: Flask Only
SDP
Catalog No. K5700010250 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
250 mL
Catalog No. Capacity (Metric)
K570001-0250 250 mL
1 options
Catalog No. K570001-0250 Supplier DWK Life Sciences Supplier No. 5700010250
Add to Cart
Add to Cart
For evaporative concentration
  • 24/40 top joint
  • 19/22 bottom joint
TRUSTED_SUSTAINABILITY
Specifications
Product Type Extraction Flask
For Use With (Equipment) Evaporative Concentrator
Capacity (Metric) 250 mL
Capacity (English) 8.45 oz.
Includes Flask
Standard Taper Joint Bottom 19/22
Standard Taper Joint Top 24/40