Learn More
DWK Life Sciences Kimble™ Kontes™ Microflex™ Microscale Kits - Sublimation Assembly
SDP
Catalog No. K7461250001 Shop All DWK Life Sciences Products
Change view
Click to view available options
Product Type:
Funnel Condenser
Catalog No. Product Type
K746125-0001 Funnel Condenser
1 options
Catalog No. K746125-0001 Supplier DWK Life Sciences Supplier No. 7461250001
Add to Cart
Add to Cart
With standard taper 14/10 joint
  • Funnel Condenser and Inlet/Vacuum Adapter (K746035-0814)
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance USP Type I and ASTM E438 Type I, Class A
For Use With (Equipment) For microscale vessels, microscale threaded standard taper 14/10 kits and sublimation assembly
Material Borosilicate Glass
Standard Taper Joint 14/10
Product Type Funnel Condenser