Learn More
DWK Life Sciences Kimble™ Kontes™Microflex™ Microscale Kits - Vigreux Distilling Column
SDP
Catalog No. K7461500000 Shop All DWK Life Sciences Products
Change view
Click to view available options
Thread Size:
20-400
Catalog No. Thread Size
K746150-0000 20-400
1 options
This item is not returnable. View return policy
Catalog No. K746150-0000 Supplier DWK Life Sciences Supplier No. 7461500000
Add to Cart
Add to Cart
This item is not returnable. View return policy
Accessory for microscale standard taper 14/10 threaded kit.
  • With GPI 20-400 thread and standard taper 14/10 joint.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance USP Type I and ASTM E438 Type I, Class A
For Use With (Equipment) Accessory for microscale standard taper 14/10 threaded kit
Material Borosilicate Glass
Thread Size 20-400
Standard Taper Joint 14/10
Product Type Vigreux Distilling Column