Learn More
DWK Life Sciences™ Kimble™ Kontes™ MIDI-VAP 4000 Ammonia and Phenol Distillation System, Complete
GSA_VASDP
Catalog No. K4794904000 Shop All DWK Life Sciences Products
Change view
Click to view available options
Material:
Borosilicate Glass
Catalog No. Material
K4794904000 Borosilicate Glass
1 options
Catalog No. K4794904000 Supplier DWK Life Sciences™ Supplier No. 4794904000
Add to Cart
Add to Cart
Request a Quote
Includes
Ten sets of glassware, heater, and pre-cut tubing
Complete system and component parts for ammonia and phenol distillation
  • Includes glassware, heater, pre-cut tubing
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Includes Ten sets of glassware, heater, and pre-cut tubing
For Use With (Application) For drinking, ground, surface and saline waters as well as domestic and industrial wastes
Type MIDI-VAP™ 4000 Ammonia and Phenol Complete System
Material Borosilicate Glass
Certifications/Compliance USP Type I and ASTM E438, Type I, Class A