Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences KIMBLE™ KONTES™ Pear-Shaped Flask for MICROFLEX™ Kit
SDP

Catalog No. K7461100005 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
K746110-0005 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K746110-0005 Supplier DWK Life Sciences Supplier No. 7461100005
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The KIMBLE™ KONTES™ Pear-Shaped Flask for MICROFLEX™ Kit is a 5mL thin-wall pear-shaped flask designed as a component of the MICROFLEX Kit system for small-scale laboratory distillation and collection applications.

  • Thin-wall construction for efficient heat transfer
  • Capacity: 5mL
  • Pear-shaped flask designed for use with the MICROFLEX Kit system
  • Component part for small-scale laboratory distillation and collection workflows
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 5 mL
Material Borosilicate Glass (3.3)
Product Type Flask
Autoclavable Autoclavable
Disposable Reusable
Graduated No
Sterility Non-sterile
Bottom Shape Pear
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.