Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences KIMBLE™ KONTES™ Powder Funnels
SDP

Catalog No. K6290000022 Shop All DWK Life Sciences Products
Change view
Click to view available options
Diameter (Metric)Top:
75 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)Top
K629000-0022 75 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K629000-0022 Supplier DWK Life Sciences Supplier No. 6290000022
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KIMBLE™ KONTES™ Powder Funnels are useful for the addition of reactants to a variety of flasks. Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.

  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
  • Standard Taper joint design for secure connection to a variety of flasks
  • Useful for the addition of reactants or powdered solids to reaction vessels
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric)Top 75 mm
Material Borosilicate Glass
Height (Metric) 150 mm
Certifications/Compliance ASTM E438, Type I, Class A
Product Type Filling Funnel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.