Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences KIMBLE™ KONTES™ PTFE Replacement Plug for Economy 3-Hole Ultra-Ware Delivery Cap
SDP

Catalog No. K9539135001 Shop All DWK Life Sciences Products
Change view
Click to view available options
Thread Size (Metric):
53 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thread Size (Metric)
K953913-5001 53 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K953913-5001 Supplier DWK Life Sciences Supplier No. 9539135001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The KIMBLE™ KONTES™ PTFE Replacement Plug for Economy 3-Hole Ultra-Ware Delivery Cap is a precision-fit accessory designed to maintain a secure, chemical-resistant seal in Ultra-Ware delivery cap assemblies.

  • PTFE construction provides excellent chemical resistance and durability
  • Designed as a direct replacement plug for the Economy 3-Hole Ultra-Ware Delivery Cap
  • 1/4-28 thread fitting ensures a secure, leak-resistant connection
  • Maintains integrity of delivery cap assembly during laboratory use
  • Supplied by DWK Life Sciences under the Kimble™ Kontes™ brand
TRUSTED_SUSTAINABILITY

Specifications

Color White
Material PTFE
No. per Case 1 Cs.
Sterility Non-sterile
Thread Size (Metric) 53 mm
Neck Finish 1/4-28
For Use With (Equipment) Three hole delivery cap system
Autoclavable Autoclavable
Disposable Reusable
Product Type Plug for Delivery Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.