Description
- Designed for use with jacketed reactors
- Center joint 45/50 and outer joint 24/40
- Manufactured from borosilicate glass
Specifications
Specifications
| Material | Glass |
| For Use With (Application) | For jacketed reactors |
| Product Type | Kontes™ reaction flask top |
| Standard Taper Joint | 45/50 Center, 24/40 side |
| Autoclavable | Autoclavable |
| Disposable | Reusable |
| Mouth Style | Joint |
| Sterility | Non-sterile |
| Quantity | 1/Cs. |