Learn More
DWK Life Sciences KIMBLE™ KONTES™ Replacement Glass Stopper for Cylindrical Addition Funnels with Pressure Equalizing Lines
Shop All DWK Life Sciences Products

Description
- Replacement glass stopper designed for cylindrical addition funnels with pressure equalizing lines
- Hollow hex head design for secure seating and easy handling
- Standard Taper joint ensures compatibility with corresponding funnel joints
Specifications
Specifications
| Shape | Hexagonal Grip |
| Material | Borosilicate Glass |
| Autoclavable | Autoclavable |
| Color | Clear |
| Diameter (Metric) Outer | 14 mm |
| Size | Medium Length |
| For Use With (Equipment) | Cylindrical addition funnels with pressure equalizing lines |
| Standard Taper Joint | 14/20 |
| Product Type | Hexagonal Head Stopper |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.