Learn More
DWK Life Sciences KIMBLE™ KONTES™ Solid Standard Taper Pennyhead Stoppers
Shop All DWK Life Sciences Products

Description
- All stoppers are solid except Standard Taper stopper sizes 32 and 38, which are hollow
- Made in accordance with ASTM Standard E694
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Specifications
| Shape | Tapered |
| Autoclavable | Autoclavable |
| Color | Clear |
| Material | Glass |
| For Use With (Equipment) | Laboratory glassware such as flasks, mixing cylinders and separatory funnels |
| Product Type | Pennyhead Stopper |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.