Learn More
DWK Life Sciences Kimble-Chase Kontes™ Hi-Vac Stirrer Accessory, Stirrer Bearing Assembly
SDP
Catalog No. K7850310124 Shop All DWK Life Sciences Products
Change view
Click to view available options
For Use With (Equipment):
Hi-Vac Stirrer
Catalog No. For Use With (Equipment)
K7850310124 Hi-Vac Stirrer
1 options
Catalog No. K7850310124 Supplier DWK Life Sciences Supplier No. 7850310124
Add to Cart
Add to Cart
Add this Kimble accessory to your Hi-Vac Stirrer when using a polished glass shaft with a PTFE blade for stirring applications.
  • For use with Hi-Vac Stirrer
  • Shaft (K7865200261) and blade (K7890300022) sold separately
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Hi-Vac Stirrer
Standard Taper Joint 24/40