Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences KIMBLE™ KONTES™ Three Neck Round Bottom Heavy Wall Flasks
SDP

Catalog No. K6060001524 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
1000 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
K6060001524 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K6060001524 Supplier DWK Life Sciences Supplier No. 6060001524
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KIMBLE™ KONTES™ Three Neck Round Bottom Heavy Wall Flasks feature standard taper outer joints and vertical side necks. Made from borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements. Referenced to ASTM Method D1744.

  • Three-neck heavy wall round-bottom flask with standard taper outer joints
  • Side necks are vertical
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
  • Referenced to ASTM Method D1744
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 1000 mL
Neck Style Vertical
No. per Case 3/Cs.
Product Type Distilling Flask
Standard Taper Joint 24/40 Center, 24/40 Side
Autoclavable Autoclavable
Disposable Reusable
Graduated No
Mouth Style Joint
Sterility Non-sterile
Bottom Shape Round
Diameter (Metric) Outer 130 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.