Learn More
Description
- Specially designed for the preparation, storage, and delivery of all liquid chromatography mobile phases
- Reservoir manufactured from Type I, Class A borosilicate glass to prevent leaching of extractables into mobile phase solvents
- PVC plastic safety coating helps retain glass fragments in case of an accident
- UV blocking agent in coating blocks virtually all UV light up to 385nm, preventing photo-degradation of light-sensitive reagents
- Side neck allows addition of filtered solvent without removing the delivery cap
- Conical bottom design allows delivery of virtually all mobile phase without dangerous reservoir tilting
- GL 45 screw thread; operating pressure range -1.0 to 0.4 bar
Specifications
Specifications
| Certifications/Compliance | USP Type I and ASTM E438, Type I, Class A |
| Coating | PVC |
| Diameter (English) Outer | 8.07 in. |
| Capacity (Metric) | 5000 mL |
| Material | Borosilicate Glass |
| Sterility | Non-sterile |
| Type | HPLC Reservoir |
| For Use With (Application) | Designed for the preparation, storage, and delivery of liquid chromatography mobile phases |
| Height (English) | 14.68 in. |
| Height (Metric) | 373 mm |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
