Learn More
DWK Life Sciences KIMBLE™ KONTES™ Ultra-Ware™ Three-Hole Cap Systems
GSA_VASDP
Catalog No. K9539135000 Shop All DWK Life Sciences Products
Change view
Click to view available options
Thread Size (Metric):
GL 45
Catalog No. Thread Size (Metric)
K953913-5000 GL 45
1 options
Catalog No. K953913-5000 Supplier DWK Life Sciences Supplier No. 9539135000
Add to Cart
Add to Cart
KIMBLE™ KONTES™ Ultra-Ware™ Three-Hole Cap Systems offer borosilicate glass and PTFE construction for excellent chemical resistance to HPLC mobile phases. Supplied with three-hole cap, conical bottom reservoir, standard fittings kit, and PTFE tubing.
Borosilicate glass and PTFE construction give excellent chemical resistance to HPLC mobile phases
  • Supplied with three-hole cap, conical bottom reservoir, standard fittings kit, and PTFE tubing
  • Reservoir manufactured from Type I, Class A borosilicate glass with a plastic safety coating that absorbs UV up to 385 nm, preserves UV-sensitive mobile-phase components
  • PTFE cap body supplied with a TFE/propylene O-ring and a polypropylene screw collar
  • Two ports have 0.125 in. through holes; third port has a 0.0625 in. through hole
  • Connecting threads on top and bottom of cap use 0.25 to 28 flangeless fittings and adapters
  • Filtration of the mobile phase directly into the reservoir requires additional purchase of a filtration cap and appropriate solvent pickup adapter
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Color Blue
Material PTFE
No. per Case 1 Cs.
Sterility Non-sterile
Thread Size (Metric) GL 45
Neck Finish 53 mm
Autoclavable Autoclavable
Disposable Reusable
Product Type Multiple Distributor Screw Cap