Learn More
DWK Life Sciences KIMBLE™ KONTES™ Valve Stems for Ultra-Ware™ THF-Resistant Filtration/Delivery Four Valve Cap System
Shop All DWK Life Sciences Products

Description
- Replacement CTFE valve stems for the Kontes Ultra-Ware THF-resistant four valve recirculation/filtration cap system
- Short valve stems designed for sparge and pump ports
- Long valve stems designed for filter, vent, and recirculate ports
- CTFE (chlorotrifluoroethylene) construction provides resistance to THF and other aggressive solvents
- Sold as individual replacement components to maintain and service the four valve cap system
Specifications
Specifications
| Color | Silver |
| Material | Stainless Steel |
| No. per Case | 1 Cs. |
| Sterility | Non-sterile |
| For Use With (Equipment) | Filter and vent port |
| Autoclavable | Autoclavable |
| Disposable | Reusable |
| Product Type | Valve Stem |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.