Learn More
DWK Life Sciences KIMBLE™ Two-Leg Lyophilization Serum Bottle and Vial Stoppers
Shop All DWK Life Sciences Products

Description
For research and pharmaceutical packaging applications
- Contain no plasticizers, 2-mercapto-benzothiazole, nitrosamine precursors, or natural rubber latex
- Applicable to aqueous solutions with a pH range of 2 to 10
- For use with 20mm serum-finished bottles or vials
- Universal gray chlorobutyl formulation meets USP, EP, and JP requirements for globally marketed pharmaceutical products
Order Info
Durometer 50.
Specifications
Specifications
| Stopper Number | 20 |
| Shape | Cylindrical |
| Sterility | Non-sterile |
| Diameter (Metric) Top | 20 mm |
| Product Type | Rubber Stopper |
| Diameter (Metric) Outer | 20 mm |
| Autoclavable | Autoclavable |
| Diameter (Metric) Bottom | 13 mm |
| Material | Butyl Rubber |
| Color | Gray |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.