Learn More
DWK Life Sciences Kimble™ Closures for 20mL Glass and Plastic Scintillation Vials
SDP
Catalog No. p-206243 Shop All DWK Life Sciences Products
Change view
Click to view available options
Material:
Polyethylene
Urea
Material (Liner):
Linerless
PTFE/Rubber
Polyethylene
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Material (Liner)
03-340-250 Polyethylene Linerless
03-337-8 Urea Polyethylene
03-340-131 Urea PTFE/Rubber
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 03-340-250 Supplier DWK Life Sciences Supplier No. 7452222400
Only null left
Add to Cart
Add to Cart
Durable screw thread closures
  • For 20mL Fisherbrand™ and Kimble scintillation vials, except 03-340-4B and 03-340-129. GPI 22-400 thread.
TRUSTED_SUSTAINABILITY
Specifications
Material (Liner) Linerless
Color White
Material Polyethylene