Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ KIMAX™ Borosilicate Petri Dish Set
SDPSCI_ED

Catalog No. S39362 Shop All DWK Life Sciences Products
Change view
Click to view available options
Diameter (Metric):
100 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
S39362 100 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S39362 Supplier DWK Life Sciences Supplier No. 2306110020/EMD

missing translation for 'validMainframeMsg'

Shallow glass dishes used for culturing bacteria and other general laboratory uses. Designed to meet Federal Specification NNN-D-1478.

  • Kimble Kimax dishes remain clear after repeated use in wet or dry sterilization cycles
  • Reinforced beaded edges resist mechanical breakage and help in centering the bottom inside the cover
  • Covers have a fused-on bright red ceramic enamel marking spot and inscription, including “Cover,” Bottoms have a white ceramic enamel inscription, including “Bottom”
  • Combination of different colors and wording permits quick and easy identification of the parts when sorting, selecting and assembling pairs
  • Bottom has a vertical arrow on the side to precisely locate serial dilution starting points
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
TRUSTED_SUSTAINABILITY

Specifications

Material Borosilicate Glass
Shape Round
Type Petri Dish Set
Height (Metric) 20 mm
Diameter (Metric) 100 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.