Learn More
DWK Life Sciences KIMBLE™ KIMAX™ Glass Buret Tip
SDP
Catalog No. 1031031 Shop All DWK Life Sciences Products
Change view
Click to view available options
Material:
PTFE
Catalog No. Material
10-310-31 PTFE
1 options
Catalog No. 10-310-31 Supplier DWK Life Sciences Supplier No. 17034H99
Add to Cart
Add to Cart
KIMBLE™ KIMAX™ Glass Buret Tip is a threaded replacement tip compatible with 03-700-12 Series and 03-857 Series burets, fitting both 25 mL and 50 mL sizes for reliable liquid dispensing.
  • Threaded glass buret tip for secure attachment
  • Compatible with 03-700-12 Series and 03-857 Series burets
  • Fits both 25 mL and 50 mL buret sizes
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 17021H and 17121H Straight Bore PTFE Stopcock Burettes
Material PTFE
For Use With (Application) Calibrated to Accurately Deliver Buret Contents at a Proper Rate
Type Buret Tip