Learn More
DWK Life Sciences KIMBLE™ GLS 80™ KimCote™ Clear Wide Mouth Media Bottles with Caps
Shop All DWK Life Sciences Products

Description
- Wide-mouth
- 3.3 borosilicate glass
- Durable ceramic ink graduations and marking patch
- Blue quick-release linerless polypropylene closure and pour ring are rated up to 140° C
- Autoclavable
- KimCote™ coating provides safety and reduces hazards of shattered glass
Specifications
Specifications
| Bottle Surface | KimCote™ |
| Capacity (Metric) | 2000 mL |
| Capacity (English) | 64 oz. |
| Coating | KimCote™ |
| Color | Clear |
| For Use With (Application) | Storage and Transport |
| Includes | Blue Polypropylene Cap and Pouring Ring |
| Material | Borosilicate Glass (3.3) |
| Length (Metric) | 292.1 mm |
| Max. Temperature (Metric) | 140°C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.